Abnova
Manufacturer Code:H00010881B01
Catalog # 89-007-075
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | ACTL7A (NP_006678.1 1 a.a. ~ 435 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human ACTL7A protein.
For Research Use Only
Protein Aliases: actin-like 7-alpha; actin-like 7A; Actin-like protein 7A; Actin-like-7-alpha; testicular secretory protein Li 3
Gene Aliases: ACTL7A
UniProt ID: (Human) Q9Y615
Entrez Gene ID: (Human) 10881
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.