Novus Biologicals
Manufacturer Code:NBP267113
Catalog # NBP267113
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JF47-01 |
Immunogen | Full length protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ACTA actin alpha 1 skeletal muscle alpha skeletal muscle alpha skeletal muscle actin alpha-actin-1 ASMA CFTD CFTDM MPFD NEM1 NEM2 NEM3; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; Alpha-actin-1; nemaline myopathy type 3
Gene Aliases: ACTA; ACTA1; ASMA; CFTD; CFTD1; CFTDM; MPFD; NEM1; NEM2; NEM3; SHPM
UniProt ID: (Human) P68133
Entrez Gene ID: (Human) 58
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.