Invitrogen
Manufacturer Code:MA511866
Catalog # PIMA511866
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ACTN05 (C4) |
Immunogen | Chicken gizzard actin |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: a-actin; actin alpha 1; actin alpha cardiac; actin alpha cardiac 1; actin alpha skeletal muscle alpha skeletal muscle actin alpha-actin-1 alpha-sma Atcb; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; Actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; Actin, cytoplasmic 1; Actin, cytoplasmic 1, N-terminally processed; actin, gamma 2, smooth muscle, enteric; Actin, gamma-enteric smooth muscle; Actin, gamma-enteric smooth muscle, intermediate form; actin-like protein; alpha actin 1; alpha actin 2; Alpha-actin; alpha-actin cardiac; Alpha-actin-1; Alpha-actin-2; Alpha-actin-3; Alpha-cardiac actin; alpha-smooth muscle actin; alphac-actin; B-actin; Bact; actin; beta actin; beta cytoskeletal actin; Beta-actin; cardiac actin; cardiac muscle alpha actin 1; Cell growth-inhibiting gene 46 protein; cytoplasmic 1; cytoplasmic beta-actin; cytoskeletal beta actin; gamma non-muscle actin; Gamma-2-actin; gamma-enteric smooth muscle actin; nemaline myopathy type 3; PS1TP5-binding protein 1; skeletal alpha actin; smooth muscle alpha-actin; Smooth muscle gamma-actin; unnamed protein product; vascular smooth muscle alpha-actin
Gene Aliases: 0610041G09Rik; a-SMA; AA959943; AAT6; ACT; ACT-4; ACT-5; Act4; ACTA; Acta-2; ACTA1; ACTA2; ACTA3; ACTB; ACTC; Actc-1; ACTC1; ACTE; ACTG2; ACTGE; actin; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; ACTVS; Actx; alphaSMA; ASD5; ASMA; Bact; beta-actin; BRWS1; CFTD; CFTD1; CFTDM; CMD1R; CMH11; E430023M04Rik; GAMMA-ACTIN; GIG46; LVNC4; MPFD; MYMY5; NEM1; NEM2; NEM3; PS1TP5BP1; SHPM; SMalphaA; SMGA; VSCM
UniProt ID: (Chicken) P68139, (Human) P68133, (Bovine) P68138, (Pig) P68137, (Rabbit) P62740, (Chicken) P08023, (Bovine) P62739, (Human) P62736, (Rabbit) P29751, (Bovine) P60712, (Pig) Q6QAQ1, (Human) P60709, (Chicken) P60706, (Dog) O18840, (Human) P68032, (Bovine) Q3ZC07, (Chicken) P68034, (Bovine) Q5E9B5, (Chicken) P63270, (Human) P63267, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P60711, (Mouse) P60710, (Rat) P68035, (Mouse) P68033, (Mouse) P63268, (Rat) P63269
Entrez Gene ID: (Chicken) 421534, (Human) 58, (Bovine) 281592, (Rabbit) 100009506, (Pig) 100154254, (Dog) 488984, (Pig) 733615, (Dog) 477587, (Rabbit) 100009271, (Chicken) 423787, (Bovine) 515610, (Human) 59, (Rabbit) 100009272, (Bovine) 280979, (Pig) 414396, (Human) 60, (Chicken) 396526, (Dog) 403580, (Dog) 478250, (Human) 70, (Bovine) 533219, (Chicken) 423298, (Rabbit) 100340339, (Pig) 100152267, (Bovine) 281595, (Chicken) 396084, (Pig) 100520667, (Human) 72, (Rabbit) 100350318, (Dog) 475792, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Rat) 81822, (Mouse) 11461, (Rat) 29275, (Mouse) 11464, (Mouse) 11468, (Rat) 25365
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.