Invitrogen
Manufacturer Code:PA578717
Catalog # PIPA578717
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human Actin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin cytoplasmic 2 Gamma-actin Actin cytoplasmic 2 N-terminally processed ACTG1 ACTG; Actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; actin, gamma, cytoplasmic 1; beta-actin FE-3; cytoskeletal gamma-actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; epididymis luminal protein 176; Gamma-actin
Gene Aliases: ACT; ACTG; ACTG1; Actl; AL023024; BRWS2; DFNA20; DFNA26; E51; HEL-176
UniProt ID: (Human) P63261, (Mouse) P63260, (Rat) P63259
Entrez Gene ID: (Human) 71, (Mouse) 11465, (Rat) 287876
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.