R&D Systems
Manufacturer Code:MAB9308100
Catalog # MAB9308100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 959606 |
Immunogen | <em>E.coli</em>-derived recombinant human alpha-Cardiac Actin Asp3-Phe377 Accession # P68032 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ACTC ACTC1 actin alpha cardiac muscle 1 ASD5 Cardiac alpha-Actin CMD1R CMH11 LVNC4; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Alpha-cardiac actin
Gene Aliases: ACTC; ACTC1; ASD5; CMD1R; CMH11; LVNC4
UniProt ID: (Human) P68032
Entrez Gene ID: (Human) 70
Molecular Function:
actin and actin related protein
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.