Novus Biologicals
Manufacturer Code:NBP197725JF646
Catalog # NBP97725646
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoblot (IB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 22D3 |
Immunogen | Clone 22D3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-cardiac actin with an acetylated N-terminus (Ac-DDEETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ACTC actin alpha cardiac muscle 1 ASD5 CMD1R CMH11 LVNC4; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; Alpha-cardiac actin
Gene Aliases: ACTC; ACTC1; ASD5; CMD1R; CMH11; LVNC4
UniProt ID: (Human) P68032
Entrez Gene ID: (Human) 70
Molecular Function: actin and actin related protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.