Invitrogen
Manufacturer Code:PA529962
Catalog # PIPA529962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 216 of Human Acrosomal vesicle protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acrosomal protein SP-10; Acrosomal vesicle protein 1; D11S4365 SP-10 SPACA2 acrosomal protein SP-10 sperm protein 10; MSA-63
Gene Aliases: Acrv1; Msa63; SP-10
UniProt ID: (Mouse) P50289
Entrez Gene ID: (Mouse) 11451
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.