Novus Biologicals
Manufacturer Code:NB500419UV
Catalog # NB500419UV
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Pig |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | ACR-2 |
Immunogen | Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization. |
Conjugate | DyLight 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acrosin; acrosin ACRS preproacrosin proacrosin; Acrosin heavy chain; acrosin light and heavy chain prepropeptide; Acrosin light chain; preproacrosin; proacrosin
Gene Aliases: ACR; ACRS
UniProt ID: (Human) P10323
Entrez Gene ID: (Human) 49
Molecular Function:
hydrolase
protease
serine protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.