Novus Biologicals
Manufacturer Code:NBP270074FR
Catalog # NBP270074FR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3G8 |
Immunogen | Full length recombinant protein of human Aconitase 2 (NP_001089) produced in HEK293T cells. |
Conjugate | DyLight 680 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Aconitase; aconitase 2, mitochondrial; Aconitase aconitase 2 mitochondrial aconitate hydratase mitochondrial ACONM Citrate hydro-lyase EC 4.2.1 EC 4.2.1.3 MGC20605 MGC33908; Aconitate hydratase, mitochondrial; Citrate hydro-lyase; epididymis secretory sperm binding protein Li 284; mitochondrial aconitase
Gene Aliases: ACO2; ACONM; HEL-S-284; ICRD; OCA8; OPA9
UniProt ID: (Human) Q99798
Entrez Gene ID: (Human) 50
Molecular Function:
dehydratase
hydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.