Invitrogen
Manufacturer Code:PA586325
Catalog # PIPA586325
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 100-150 of Human IRP-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aconitase; aconitase 1, soluble; aconitate hydratase, cytoplasmic; Citrate hydro-lyase; cytoplasmic aconitase; Cytoplasmic aconitate hydratase; Cytoplasmic aconitate hydratase Aconitase Citrate hydro-lyase Ferritin repressor protein Iron regulatory protein 1 IRP1 Iron-responsive element-binding protein 1 IRE-BP 1 ACO1 IREB1; cytosplasmic aconitase; epididymis luminal protein 60; Ferritin repressor protein; IRE-BP 1; Iron regulatory protein 1; iron-responsive element binding protein 1; Iron-responsive element-binding protein 1; IRP1; soluble aconitase
Gene Aliases: Aco-1; ACO1; Acon1; ACONS; AH; HEL60; IREB1; IREBP; IREBP1; IRP1
UniProt ID: (Human) P21399, (Rat) Q63270, (Mouse) P28271
Entrez Gene ID: (Human) 48, (Rat) 50655, (Mouse) 11428
Molecular Function:
dehydratase
hydratase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.