Invitrogen
Manufacturer Code:PA130864
Catalog # PIPA130864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein (Human). HCPTPA (GB Accession No.M83653) was subcloned into pRSET to express a His6 N terminal fusion protein. This protein was expressed in E. coli and purified to homogeneity on Ni affinity matrix. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: acid phosphatase 1, soluble; acid phosphatase of erythrocyte; Acp1-PA; Adipocyte acid phosphatase; Adult cuticle protein 1; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; dACP-1; LMW-PTP; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; LMW-PTPase; Low molecular weight cytosolic acid phosphatase; Low molecular weight phosphotyrosine protein phosphatase; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37
Gene Aliases: 4632432E04Rik; Acp-1; ACP1; AI427468; CG7216; CG7216-PA; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; LMW-PTP; LMW-PTPase; RCJMB04_11a4
UniProt ID: (Bovine) P11064, (Chicken) Q5ZKG5, (Pig) P81693, (Human) P24666, (Rat) P41498, (Fruit fly) Q26416, (Mouse) Q9D358
Entrez Gene ID: (Bovine) 280977, (Chicken) 421909, (Pig) 100737301, (Human) 52, (Rat) 24161, (Fruit fly) 34052, (Mouse) 11431
Molecular Function:
hydrolase
oxidoreductase
phosphatase
protein phosphatase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.