Invitrogen
Manufacturer Code:PA126888
Catalog # PIPA126888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Bovine |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Acetyl Cholinesterase from bovine erythrocytes. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholinesterase; acetylcholinesterase T-subunit; AChE
Gene Aliases: ACHE
UniProt ID: (Bovine) P23795
Entrez Gene ID: (Bovine) 540446
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.