Proteintech
Manufacturer Code:179751AP150UL
Catalog # 50-173-1911
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Acetylcholinesterase Fusion Protein Ag12146 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholinesterase; acetylcholinesterase (Yt blood group); Acetylcholinesterase ACHE ARACHE N ACHE YT; AChE; apoptosis-related acetylcholinesterase; glycolipid-anchored form of acetylcholinesterase; Yt blood group
Gene Aliases: ACEE; ACHE; ARACHE; N-ACHE; YT
UniProt ID: (Human) P22303, (Mouse) P21836, (Rat) P37136
Entrez Gene ID: (Human) 43, (Mouse) 11423, (Rat) 83817
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.