R&D Systems
Manufacturer Code:MAB7574
Catalog # MAB7574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 872044 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human ACHE Met1-Leu614 Accession # P22303 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Acetylcholinesterase; acetylcholinesterase (Yt blood group); acetylcholinesterase acetylcholinesterase (Yt blood group) AChE apoptosis-related acetylcholinesterase ARACHE EC 3.1.1 EC 3.1.1.7 N-ACHE YT Yt blood group; AChE; apoptosis-related acetylcholinesterase; Yt blood group
Gene Aliases: ACEE; ACHE; ARACHE; N-ACHE; YT
UniProt ID: (Human) P22303
Entrez Gene ID: (Human) 43
Molecular Function: esterase hydrolase lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.