Novus Biologicals
Manufacturer Code:NBP105139AF750
Catalog # NP10139A70
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 12B4 |
Immunogen | Brain acetylcholinesterase human purified and detergent solubilized (1). |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Acetylcholinesterase; acetylcholinesterase (Yt blood group); acetylcholinesterase acetylcholinesterase (Yt blood group) AChE apoptosis-related acetylcholinesterase ARACHE EC 3.1.1 EC 3.1.1.7 N-ACHE YT Yt blood group; AChE; apoptosis-related acetylcholinesterase; Yt blood group
Gene Aliases: ACEE; ACHE; ARACHE; N-ACHE; YT
UniProt ID: (Human) P22303
Entrez Gene ID: (Human) 43
Molecular Function:
esterase
hydrolase
lipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.