Novus Biologicals
Manufacturer Code:NBP267545
Catalog # NBP267545
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ST53-08 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ACC-beta; ACC2ACC-beta ACCBacetyl-CoA carboxylase 2 acetyl-CoA carboxylase beta acetyl-Coenzyme A carboxylase beta EC 6.4.1.2 HACC275; Acetyl-CoA carboxylase 2; acetyl-Coenzyme A carboxylase beta
Gene Aliases: ACACB; ACC2; ACCB; HACC275
UniProt ID: (Human) O00763
Entrez Gene ID: (Human) 32
Molecular Function:
ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.