Invitrogen
Manufacturer Code:MA524698
Catalog # PIMA524698
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | RM235 |
Immunogen | Acetyl-peptide corresponding to Acetyl-Histone H2B (Lys20). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: H2B histone family member F H2B/f H2B/I H2BFA H2BFH H2BFL H2BK5ac HIST1H2BC histone 1 H2bb Histone H2B type 1-B Histone H2B.1 Histone H2B.f Histone H2B.I; H2B histone family, member A; H2B histone family, member K; H2B histone family, member Q; H2B-clustered histone 18; H2B-clustered histone 21; H2B/a; H2B/g; H2B/h; H2B/k; H2B/l; H2B/q; histone 1, H2bg; histone 1, H2bi; histone 2, H2be; Histone H2B type 1-C/E/F/G/I; Histone H2B type 2-E; Histone H2B type 2-F; Histone H2B-GL105; Histone H2B.1 A; Histone H2B.a; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l; Histone H2B.q
Gene Aliases: dJ221C16.8; GL105; H2B; H2B.1; H2B.1A; H2B/a; H2B/k; H2BC10; H2BC18; H2BC21; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFG; H2BFH; H2BFK; H2BFL; H2BFQ; H2BGL105; H2BQ; HIST1H2BC; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; HIST2H2BE; HIST2H2BF
UniProt ID: (Human) P62807, (Human) Q16778, (Human) Q5QNW6
Entrez Gene ID: (Human) 8339, (Human) 8346, (Human) 8349, (Human) 440689
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.