Invitrogen
Manufacturer Code:PA585530
Catalog # PIPA585530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Dot blot (DB) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human Histone H2BK5ac (acetyl Lys5). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H2B histone family, member U, (testis-specific); histone 1, H2ba; Histone H2B type 1-A; Histone H2B, testis; Testis-specific histone H2B; TSH2B.1
Gene Aliases: bA317E16.3; H2BC1; H2BFU; HIST1H2BA; STBP; TH2B; TSH2B; TSH2B.1
UniProt ID: (Human) Q96A08
Entrez Gene ID: (Human) 255626
Molecular Function: DNA binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.