Invitrogen
Manufacturer Code:PA516185
Catalog # PIPA516185
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ChIP assay (ChIP) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic acetylated peptide corresponding to residues surrounding Lys1535 of human CBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CREB-binding protein; Crebbp Ep300 CBP RSTS KAT3A CREBBP; E1A-associated protein p300; E1A-binding protein, 300kD; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; Histone lysine acetyltransferase CREBBP; p300 HAT; Protein 2-hydroxyisobutyryltransferase p300; Protein lactyltransferas p300; Protein propionyltransferase p300; Protein-lysine acetyltransferase CREBBP
Gene Aliases: A430090G16; A730011L11; AW558298; CBP; CBP/p300; CREBBP; EP300; KAT3A; KAT3B; P300; p300 HAT; p300/CBP; RSTS; RSTS2; RTS
UniProt ID: (Human) Q92793, (Human) Q09472, (Mouse) B2RWS6
Entrez Gene ID: (Human) 1387, (Mouse) 12914, (Rat) 54244, (Human) 2033, (Mouse) 328572, (Rat) 170915
Molecular Function: DNA binding protein acetyltransferase chromatin/chromatin-binding protein nucleic acid binding transcription cofactor transcription factor transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.