Abnova
Manufacturer Code:MAB18426
Catalog # 89-338-351
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S271-44 |
Immunogen | Recombinant protein corresponding to amino acids 460-526 at C-terminus of mouse Accn2. |
Conjugate | A594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acid sensing ion channel; Acid-sensing ion channel; Acid-sensing ion channel 1; AI843610; Amiloride-sensitive cation channel 2, neuronal; ASIC; ASIC1; ASIC1a; B530003N02Rik; BNaC2; Brain sodium channel 2; degenerin 2
Gene Aliases: Accn2; AI843610; ASIC; Asic1; ASIC1a; B530003N02Rik; BNaC2
UniProt ID: (Mouse) Q6NXK8
Entrez Gene ID: (Mouse) 11419
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.