Abnova
Manufacturer Code:MAB3780
Catalog # 89-111-590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | BXP-9 |
Immunogen | Recombinant MBP fusion protein corresponding to amino acids 221-394 of mouse Abcg2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against partial recombinant Abcg2.
For Research Use Only
Protein Aliases: ABC15; ABCP; AI428558; ATP-binding cassette sub-family G member 2; BCRP; Bcrp1; Breast cancer resistance protein 1 homolog; Broad substrate specificity ATP-binding cassette transporter ABCG2; CD338; mitoxantrone resistance protein 1; MXR; MXR1; Urate exporter
Gene Aliases: ABC15; Abcg2; ABCP; AI428558; BCRP; Bcrp1; MXR; MXR1
UniProt ID: (Mouse) Q7TMS5
Entrez Gene ID: (Mouse) 26357
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.