Abnova
Manufacturer Code:MAB18459
Catalog # 89-338-384
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S319A-14 |
Immunogen | Recombinant protein corresponding to amino acids 1505-1546 at C-terminus of mouse Abcc9. |
Conjugate | A488 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI414027; AI449286; ATP-binding cassette sub-family C member 9; ATP-binding cassette transporter sub-family C member 9; Sulfonylurea receptor 2; sulfonylurea-binding protein 2; Sur2; SUR2A; SUR2B
Gene Aliases: Abcc9; AI414027; AI449286; Sur2; SUR2A; SUR2B
UniProt ID: (Mouse) P70170
Entrez Gene ID: (Mouse) 20928
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.