Abnova
Manufacturer Code:MAB6671
Catalog # 89-112-996
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | M5I-10 |
Immunogen | Recombinant protein corresponding to amino acids 1-38 of mouse Abcc5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rat monoclonal antibody raised against partial recombinant Abcc5.
For Research Use Only
Protein Aliases: 2900011L11Rik; Abcc5a; Abcc5b; AI132311; ATP-binding cassette sub-family C member 5; ATP-binding cassette, sub-family C (CFTR/MRP), member 5a; ATP-binding cassette, sub-family C (CFTR/MRP), member 5b; ATP-binding cassette, sub-family C, member 5; MGC116567; MOAT-C; Mrp5; Multi-specific organic anion transporter C; Multidrug resistance-associated protein 5; SMRP
Gene Aliases: 2900011L11Rik; Abcc5; Abcc5a; Abcc5b; AI132311; Mrp5
UniProt ID: (Mouse) Q9R1X5
Entrez Gene ID: (Mouse) 27416
Molecular Function: ATP-binding cassette (ABC) transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.