Abnova
Manufacturer Code:PAB5860
Catalog # 89-131-954
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Abcc2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Abcc2.
For Research Use Only
Protein Aliases: Abc30; AI173996; ATP-binding cassette sub-family C member 2; Canalicular multispecific organic anion transporter 1; Cmoat; cMRP; Mrp2; multidrug resistance protein 2; Multidrug resistance-associated protein 2
Gene Aliases: Abc30; Abcc2; AI173996; Cmoat; cMRP; Mrp2
UniProt ID: (Mouse) Q8VI47
Entrez Gene ID: (Mouse) 12780
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.