Abnova
Manufacturer Code:MAB2418
Catalog # 89-110-999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 7A1-144 |
Immunogen | Recombinant protein corresponding extracellular region of mouse Abca7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rat monoclonal antibody raised against recombinant Abca7.
For Research Use Only
Protein Aliases: Abc51; ABCX; ATP-binding cassette sub-family A member 7
Gene Aliases: Abc51; Abca7; ABCX
UniProt ID: (Mouse) Q91V24
Entrez Gene ID: (Mouse) 27403
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.