Abnova
Manufacturer Code:PAB15854
Catalog # 89-117-916
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 293 amino acids of mouse Abca5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Abca5.
For Research Use Only
Protein Aliases: ABC13; ATP-binding cassette sub-family A member 5; ATP-binding cassette transporter sub-family A member 5; B930033A02Rik; Cholesterol transporter ABCA5; mKIAA1888
Gene Aliases: ABC13; Abca5; B930033A02Rik; Kiaa1888; mKIAA1888
UniProt ID: (Mouse) Q8K448
Entrez Gene ID: (Mouse) 217265
Molecular Function:
ATP-binding cassette (ABC) transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.