OriGene
Manufacturer Code:TA810908
Catalog # 50-168-3706
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5C7 |
Immunogen | Human recombit protein fragment corresponding to amino acids 260-448 of human AZIN1 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antizyme inhibitor 1; AZI; AZI AZI1 AZIA1 OAZI OAZIN ODC1L; Ornithine decarboxylase antizyme inhibitor
Gene Aliases: AZI; AZIA1; AZIN1; OAZI; OAZIN; ODC1L
UniProt ID: (Human) O14977
Entrez Gene ID: (Human) 51582, (Rat) 58961
Molecular Function: decarboxylase lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.