Invitrogen
Manufacturer Code:PA575645
Catalog # PIPA575645
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human AZGP1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-2 - glycoprotein 1 zinc; alpha-2 - glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc; Alpha-2-glycoprotein, zinc; major histocompatibility complex class I-like protein; testicular tissue protein Li 227; Zinc-alpha-2-glycoprotein; Zinc-alpha-2-glycoprotein Zn-alpha-2-GP Zn-alpha-2-glycoprotein AZGP1 ZAG ZNGP1; Zn - alpha2 - glycoprotein; zn-alpha-2-glycoprotein; Zn-alpha-2-GP; Zn-alpha2-glycoprotein
Gene Aliases: AZGP1; ZA2G; ZA2GA; ZAG; Zna2gp; ZNGP1
UniProt ID: (Human) P25311, (Mouse) Q64726
Entrez Gene ID: (Human) 563, (Mouse) 12007, (Rat) 25294
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.