Novus Biologicals
Manufacturer Code:NBP156324
Catalog # NBP156324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to AXUD1 (cysteine-serine-rich nuclear protein 1) The peptide sequence was selected from the middle region of AXUD1)(50ug). Peptide sequence ARVQTHFIHTLTRLQLEQEAESFRELEAPAQGSPPSPGEEALVPTFPLAK. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axin-1 up-regulated gene 1 protein; AXIN1 up-regulated 1; AXIN1 up-regulated 1 Axin-1 up-regulated gene 1 protein AXUD1DKFZp566F164 CSRNP-1 cysteine/serine-rich nuclear protein 1 cysteine-serine-rich nuclear protein 1 FAM130B Protein URAX1 TAIP3 TAIP-3TGF-beta-induced apoptosis protein 3 TGF-beta induced apoptosis protein 3 URAX1; CSRNP-1; Cysteine/serine-rich nuclear protein 1; Protein URAX1; TAIP-3; TGF-beta-induced apoptosis protein 3
Gene Aliases: AXUD1; CSRNP-1; CSRNP1; FAM130B; TAIP-3; TAIP3; URAX1
UniProt ID: (Human) Q96S65
Entrez Gene ID: (Human) 64651
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.