Proteintech
Manufacturer Code:131961AP150UL
Catalog # 50-172-8562
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AXL Fusion Protein Ag3898 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adhesion-related kinase; AXL AXL oncogene AXL receptor tyrosine kinase JTK11 UFO; AXL oncogene; AXL transforming sequence/gene; Tyrosine-protein kinase receptor UFO; ufo oncogene homolog
Gene Aliases: AI323647; ARK; AXL; JTK11; Tyro7; UFO
UniProt ID: (Human) P30530, (Mouse) Q00993
Entrez Gene ID: (Human) 558, (Mouse) 26362
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.