Invitrogen
Manufacturer Code:PA578247
Catalog # PIPA578247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the center region of human Axin 2. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axil; AXIL DKFZp781B0869 MGC10366 MGC126582 AXIN2 axin-like protein axin-2 axis inhibition protein 2 conductin axin 2; Axin-2; Axin-like protein; Axis inhibition protein 2; Conductin
Gene Aliases: Axi1; AXIL; AXIN2; Conductin; ODCRCS
UniProt ID: (Human) Q9Y2T1, (Mouse) O88566, (Rat) O70240
Entrez Gene ID: (Human) 8313, (Mouse) 12006, (Rat) 29134
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.