Invitrogen
Manufacturer Code:PA521093
Catalog # PIPA521093
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 20 amino acid peptide near the carboxy terminus of human AXIN2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axil; AXIL axin-2 axin-like protein axis inhibition protein 2 conductin; Axin-2; Axin-like protein; Axis inhibition protein 2; Conductin
Gene Aliases: Axi1; AXIL; AXIN2; Conductin; ODCRCS
UniProt ID: (Human) Q9Y2T1, (Mouse) O88566, (Rat) O70240
Entrez Gene ID: (Human) 8313, (Mouse) 12006, (Rat) 29134
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.