Proteintech
Manufacturer Code:205401AP150UL
Catalog # 50-173-2947
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | AXIN2 Fusion Protein Ag14574 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Axil; AXIL axin 2 Axin like protein AXIN2 Axis inhibition protein 2 Conductin; Axin-2; Axin-like protein; Axis inhibition protein 2; Conductin
Gene Aliases: Axi1; AXIL; AXIN2; Conductin; ODCRCS
UniProt ID: (Human) Q9Y2T1, (Mouse) O88566, (Rat) O70240
Entrez Gene ID: (Human) 8313, (Mouse) 12006, (Rat) 29134
Molecular Function:
G-protein modulator
enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.