Invitrogen
Manufacturer Code:PA518024
Catalog # PIPA518024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (FEEKIIGKVEKVD) corresponding to the C-terminus amino acids of AXIN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Axin-1; AXIN1 axin 1 AXIN LA16c-314G4.3 MGC52315 axis inhibition protein 1 axis inhibitor 1 fused mouse homolog of; Axis inhibition protein 1; axis inhibitor 1; fused, mouse, homolog of; hAxin; protein phosphatase 1, regulatory subunit 49
Gene Aliases: AXIN; AXIN1; PPP1R49
UniProt ID: (Human) O15169
Entrez Gene ID: (Human) 8312
Molecular Function: G-protein modulator enzyme modulator
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.