Invitrogen
Manufacturer Code:PA524313
Catalog # PIPA524313
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 303-331 amino acids from the C-terminal region of human AWAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 11-cis-RE-synthase; 11-cis-specific retinyl-ester synthase; Acyl-CoA retinol O-fatty-acyltransferase; Acyl-CoA wax alcohol acyltransferase 2; ARAT; DC4 DGAT2L4 MFAT WS diacylglycerol O-acyltransferase 2-like protein 4 diacylglycerol O-acyltransferase candidate 4 long-chain-alcohol O-fatty-acyltransferase 2 multifunctional O-acyltransferase; Diacylglycerol O-acyltransferase 2-like protein 4; Diacylglycerol O-acyltransferase candidate 4; hDC4; hWS; Long-chain-alcohol O-fatty-acyltransferase 2; Multifunctional O-acyltransferase; mWS; retinol O-fatty-acyltransferase; Wax synthase; wax synthase 2
Gene Aliases: 9430062J17Rik; ARAT; AWAT2; DC4; DGAT2L4; MFAT; WS
UniProt ID: (Human) Q6E213, (Mouse) Q6E1M8
Entrez Gene ID: (Human) 158835, (Mouse) 245532
Molecular Function:
acyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.