Invitrogen
Manufacturer Code:PA577763
Catalog # PIPA577763
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide RHTTHSLGPQDESC corresponding to amino acid residues 345-358 of rat Vasopressin V2 Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor; Arginine Vasopressin Receptor 2 Antidiuretic Hormone Receptor Nephrogenic Diabetes Insipidus V2R; AVPR V2; nephrogenic diabetes insipidus; Renal-type arginine vasopressin receptor; V2R; Vasopressin receptor V2; Vasopressin V2 receptor
Gene Aliases: ADHR; Avpr2; DI1; DIR; ND1; V2R; VPV2R
UniProt ID: (Mouse) O88721, (Rat) Q00788
Entrez Gene ID: (Mouse) 12000, (Rat) 25108
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.