Invitrogen
Manufacturer Code:PA572221
Catalog # PIPA572221
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 343-377 amino acids from the C-terminal region of human AVPR2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor; AVPR V2; AVPR2 ADHR AVPR V2 Avp2r Antidiuretic hormone receptor DI1 DIR3 NDI Vasopressin type 2 receptor Vasopressin type II receptor Vasopressin V2 receptor V2 vasopressin receptor Vasopressin receptor type 2 DIR Type 2 vasopressin receptor V2-renal V2R V2R; Renal-type arginine vasopressin receptor; V2R; Vasopressin V2 receptor
Gene Aliases: ADHR; AVPR2; DI1; DIR; DIR3; NDI; V2R
UniProt ID: (Human) P30518
Entrez Gene ID: (Human) 554
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.