Invitrogen
Manufacturer Code:PA120644
Catalog # PIPA120644
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide (aa sequence is considered to be commercially sensitive). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADHR DI1 DIR DIR3 MGC126533 MGC138386 NDI V2R; Antidiuretic hormone receptor; AVPR V2; Renal-type arginine vasopressin receptor; V2R; Vasopressin V2 receptor
Gene Aliases: ADHR; AVPR2; DI1; DIR; DIR3; NDI; V2R
UniProt ID: (Human) P30518
Entrez Gene ID: (Human) 554
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.