Invitrogen
Manufacturer Code:PA577762
Catalog # PIPA577762
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)RAKPRPAGSLKDLE corresponding to amino acid residues 399-412 of rat Vasopressin V1B Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1b; arginine vasopressin type 1b receptor; AVPR V1b; AVPR V1b AVPR V3 Antidiuretic hormone receptor 1b Vasopressin V3 receptor; AVPR V3; V1bR; V3/V1b pituitary vasopressin receptor; Vasopressin V1b receptor; Vasopressin V3 receptor
Gene Aliases: Avpr1b; AVPR3; V1br; V3/V1b; VIBR; VPR3
UniProt ID: (Mouse) Q9WU02
Entrez Gene ID: (Mouse) 26361, (Rat) 29462
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.