Invitrogen
Manufacturer Code:PA575375
Catalog # PIPA575375
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to amino acids 300-350 of Human V1b Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1b; arginine vasopressin receptor 3; arginine vasopressin type 1b receptor; AVPR V1b; AVPR V3; pituitary vasopressin receptor 3; V1bR; V3/V1b pituitary vasopressin receptor; Vasopressin V1b receptor; Vasopressin V1b receptor V1bR AVPR V1b AVPR V3 Antidiuretic hormone receptor 1b Vasopressin V3 receptor AVPR1B AVPR3 VPR3; Vasopressin V3 receptor
Gene Aliases: AVPR1B; AVPR3; V1br; V3/V1b; VIBR; VPR3
UniProt ID: (Human) P47901, (Mouse) Q9WU02
Entrez Gene ID: (Human) 553, (Mouse) 26361, (Rat) 29462
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.