Invitrogen
Manufacturer Code:PA533348
Catalog # PIPA533348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from C-terminus of human AVPR1B. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1b; arginine vasopressin receptor 3; AVPR V1b; AVPR V3; AVPR1B AVPR3 V1bR AVPR V1b AVPR V3 V1b receptor V1b vasopressin receptor Vasopressin V1b receptor Vasopressin V3 receptor VPR3; pituitary vasopressin receptor 3; V1bR; Vasopressin V1b receptor; Vasopressin V3 receptor
Gene Aliases: AVPR1B; AVPR3; VPR3
UniProt ID: (Human) P47901
Entrez Gene ID: (Human) 553
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.