Invitrogen
Manufacturer Code:MA518115
Catalog # PIMA518115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 721CT25.2.1 |
Immunogen | KLH conjugated synthetic peptide selected from the 353-383 region of human AVPR1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1a; AVPR V1a; AVPR1AAVPR1 Vasopressin V1a receptor Vasopressin V1a receptor AVPR V1a Vasopressin V1a receptor Antidiuretic hormone receptor 1a Vasopressin V1a receptor Vascular/hepatic-type arginine vasopressin receptor; SCCL vasopressin subtype 1a receptor; V1-vascular vasopressin receptor AVPR1A; V1a arginine vasopressin receptor; V1a vasopressin receptor; V1aR; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor
Gene Aliases: AVPR; AVPR V1a; AVPR1; AVPR1A; V1a; V1aR
UniProt ID: (Human) P37288, (Mouse) Q62463
Entrez Gene ID: (Human) 552, (Mouse) 54140
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.