Invitrogen
Manufacturer Code:PA518665
Catalog # PIPA518665
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (CHPLKTLQQPARRSR) corresponding to the internal amino acids of AVPR1A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1a; arginine vasopressin receptor V1a; AVPR V1a; AVPR1A arginine vasopressin receptor 1A AVPR1 SCCL vasopressin subtype 1a receptor V1-vascular vasopressin receptor AVPR1A V1a vasopressin receptor antidiuretic hormone receptor 1A vascular/hepatic-type arginine vasopressin receptor; SCCL vasopressin subtype 1a receptor; V1-vascular vasopressin receptor AVPR1A; V1a arginine vasopressin receptor; V1a vasopressin receptor; V1aR; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor
Gene Aliases: AVPR; AVPR V1a; AVPR1; AVPR1A; V1a; V1aR
UniProt ID: (Human) P37288, (Rat) P30560, (Mouse) Q62463
Entrez Gene ID: (Human) 552, (Rat) 25107, (Mouse) 54140
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.