Abnova
Manufacturer Code:H00000552A01
Catalog # 89-104-133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. to 52 a.a) partial recombinant protein with GST tag.,AVPR1A (NP_000697 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant AVPR1A.
For Research Use Only
Protein Aliases: Antidiuretic hormone receptor 1a; AVPR V1a; AVPR1; SCCL vasopressin subtype 1a receptor; V1-vascular vasopressin receptor AVPR1A; V1a vasopressin receptor; V1aR; Vascular/hepatic-type arginine vasopressin receptor; Vasopressin V1a receptor
Gene Aliases: AVPR V1a; AVPR1; AVPR1A; V1aR
UniProt ID: (Human) P37288
Entrez Gene ID: (Human) 552
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.