Invitrogen
Manufacturer Code:PA5112845
Catalog # PIPA5112845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Vasopressin-neurophysin 2-copeptin protein (126-164AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADH antidiuretic hormone arginine vasopressin arginine vasopressin (Diabetes insipidus) same as Di (conflicting physical mapping) arginine vasopressin (neurophysin II antidiuretic hormone diabetes insipidus neurohypophyseal) arginine vasopressin-neurophysin II precursor Arginine-vasopressin Arg-vasopressin ARVP Avp AVP-neurophysin II precursor AVP-NPII AVRP Copeptin DI neurohypophyseal Neurophysin 2 Neurophysin-I Neurophysin-II prepro-arginine-vasopressin-neurophysin II prepro-AVP-NP II prepropressophysin preprovasopressin-neurophysin Vas Vasopressin-neurophysin 2-copeptin vasopressin-neurophysin II-copeptin vasopressin-neurophysin prepropeptide VP; antidiuretic hormone; Arg-vasopressin; arginine vasopressin (Diabetes insipidus), same as Di (conflicting physical mapping); Arginine-vasopressin; AVP-NPII; Copeptin; neurohypophyseal; Neurophysin 2; Neurophysin-I; Neurophysin-II; prepro-arginine-vasopressin-neurophysin II; prepro-AVP-NP II; prepropressophysin; preprovasopressin-neurophysin; Vasopressin-neurophysin 2-copeptin; vasopressin-neurophysin II-copeptin; vasopressin-neurophysin prepropeptide
Gene Aliases: ADH; ARVP; AVP; AVP-NPII; AVRP; DI; Vas; VP
UniProt ID: (Human) P01185, (Rat) P01186
Entrez Gene ID: (Human) 551, (Rat) 24221
Molecular Function:
neuropeptide
peptide hormone
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.