Invitrogen
Manufacturer Code:PA568521
Catalog # PIPA568521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human AVIL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-binding protein DOC6; ADVIL DOC6 p92; Advillin; Advillin-like protein; p92; Peripheral nervous system villin-like protein; Pervin
Gene Aliases: ADVIL; AVIL; DOC6; p92; Pervin; zgc:136857
UniProt ID: (Human) O75366, (Mouse) O88398, (Rat) Q9WU06
Entrez Gene ID: (Bovine) 520771, (Human) 10677, (Rabbit) 100341728, (Dog) 474420, (Horse) 100051151, (Mouse) 11567, (Rat) 79253, (Zebrafish) 678618
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.