Novus Biologicals
Manufacturer Code:NBP276530
Catalog # NBP276530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL7084 |
Immunogen | This antibody was developed against a synthetic peptide corresponding to amino acids: GISPLPGGERFPYPSFHWD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: autism susceptibility candidate 2 autism susceptibility gene 2 protein autism-related protein 1 KIAA0442FBRSL2 MGC13140; Autism susceptibility gene 2 protein; autism-related protein 1
Gene Aliases: AUTS2; FBRSL2; KIAA0442; MRD26
UniProt ID: (Human) Q8WXX7
Entrez Gene ID: (Human) 26053
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.