Invitrogen
Manufacturer Code:MA531446
Catalog # PIMA531446
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | CL7067 |
Immunogen | Synthetic peptide corresponding to Human AUTS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Autism susceptibility gene 2 protein; autism-related protein 1; FBRSL2 KIAA0442
Gene Aliases: 2700063G02Rik; A730011F23Rik; AUTS2; D830032G16Rik; FBRSL2; KIAA0442; MRD26
UniProt ID: (Human) Q8WXX7
Entrez Gene ID: (Human) 26053, (Mouse) 319974
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.