Invitrogen
Manufacturer Code:PA541750
Catalog # PIPA541750
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human AUH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-methylglutaconyl-CoA hydratase; AU RNA binding protein/enoyl-CoA hydratase; AU RNA binding protein/enoyl-Coenzyme A hydratase; AU RNA-binding protein/enoyl-Coenzyme A hydratase; AU-binding enoyl-CoA hydratase; AU-binding protein/Enoyl-CoA hydratase; AU-specific RNA-binding enoyl-CoA hydratase; Itaconyl-CoA hydratase; Methylglutaconyl-CoA hydratase, mitochondrial; muAUH
Gene Aliases: AUH; C77140; W91705
UniProt ID: (Human) Q13825, (Rat) F1LU71, (Mouse) Q9JLZ3
Entrez Gene ID: (Human) 549, (Horse) 100064392, (Dog) 476348, (Rabbit) 108176743, (Bovine) 614716, (Rat) 361215, (Mouse) 11992
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.