Novus Biologicals
Manufacturer Code:NBP297719AF594
Catalog # NB103133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human AUH fragment. |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-methylglutaconyl-CoA hydratase; 3-methylglutaconyl-CoA hydratase AU RNA binding protein/enoyl-CoA hydratase AU RNA binding protein/enoyl-Coenzyme A hydratase AU RNA-binding protein/enoyl-Coenzyme A hydratase AU-binding protein/enoyl-CoA hydratase AU-specific RNA-binding enoyl-CoA hydratase EC 4.2.1.18 methylglutaconyl-CoA hydratase mitochondrial; AU RNA-binding protein/enoyl-Coenzyme A hydratase; AU-binding protein/Enoyl-CoA hydratase; AU-specific RNA-binding enoyl-CoA hydratase; Itaconyl-CoA hydratase; Methylglutaconyl-CoA hydratase, mitochondrial
Gene Aliases: AUH
UniProt ID: (Human) Q13825
Entrez Gene ID: (Human) 549
Molecular Function:
acetyltransferase
acyltransferase
dehydrogenase
epimerase/racemase
hydratase
isomerase
ligase
lyase
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.